L(+)-Cystein Hydrochlorid

Lieferant: Thermo Fisher Scientific
Achtung

111780050 111781000 111785000
ACRO111780050EA 24.4 EUR
ACRO111780050 ACRO111781000 ACRO111785000
L(+)-Cystein Hydrochlorid
L(+)-Cystein Hydrochlorid
Formel: HSCH₂CH(NH₂)COOH·HCl
Schmelzpunkt: 175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064553
CAS-Nummer: 52-89-1
EINECS: 200-157-7
Merck Index: 13,02810

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR