Dibenzosuberenon 97%
Lieferant: Thermo Scientific
Synonyme:
Dibenzo[b,f]cyclohepten-1-on, 1-Dibenzo[b,f]cycloheptenon, 5-Dibenzosuberenon
112531000
ACRO112531000EA
144
EUR
ACRO112531000
Dibenzosuberenon 97%
Dibenzosuberenon
Formel:
C₁₅H₁₀O Molecular Weight: 206,24 g/mol Siedepunkt: 347 °C (1013 hPa) Schmelzpunkt: 85…91 °C |
MDL:
MFCD00003588 CAS-Nummer: 2222-33-5 EINECS: 233-774-5 |
Spezifikation Testergebnisse
Appearance | White to yellow to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 85 to 91 °C |
Loss on drying | ≤0.5 % (60 °C) (vacuum) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...