3,4-Dichloranilin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

113110010 113110050 113112500
ACRO113110010EA 182 EUR
ACRO113110010 ACRO113110050 ACRO113112500
3,4-Dichloranilin 98%
3,4-Dichloranilin
Formel: C₆H₅Cl₂N
Molecular Weight: 162.02 g/mol
Siedepunkt: 272
Schmelzpunkt: 69…73 °C
Flammpunkt: 166 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007768
CAS-Nummer: 95-76-1
EINECS: 202-448-4
UN: 3442
ADR: 6.1 II
Merck Index: 13,03079

Bestellen

Spezifikation Testergebnisse

Appearance White to grey to brown Crystals or flakes or crystalline mass and/or chunks (May darken on storage)
Infrared spectrum Conforms
Melting point 70 to 74 °C
GC ≥97.5 %
Specific optical rotation On anhydrous basis
Titration with HClO4 97.5 to 102.5 % (On anhydrous basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR