1,2-Dichlorbenzol 99%, rein
Lieferant: Thermo Fisher Scientific
|
Achtung
|
113180010
113180025
113180250
ACRO113180010EA
48.3
EUR
ACRO113180010
ACRO113180025
ACRO113180250
1,2-Dichlorbenzol 99%, rein
1,2-Dichlorbenzol
Formel:
C₆H₄Cl₂ Siedepunkt: 179…180 °C (760 mmHg) Schmelzpunkt: –15 °C Dichte: 1,3 g/cm³ Flammpunkt: 67 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000535 CAS-Nummer: 95-50-1 EINECS: 202-425-9 UN: 1591 ADR: 6.1,III Merck Index: 13,03081 |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5500 to 1.5520 (20°C, 589 nm) |
GC | ≥98.5 % |
Suitability | for use as a laser dye |
Molar extinction coefficient | ≥105000 (lambda max) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...