2,4-Dichlorbenzylamin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: (2,4-Dichlorophenyl)methanamine

113230100 113230500
ACRO113230100EA 37.6 EUR
ACRO113230100 ACRO113230500
2,4-Dichlorbenzylamin 98%
2,4-Dichlorbenzylamin
Formel: C₇H₇Cl₂N
Molecular Weight: 176.04 g/mol
Siedepunkt: 83
Dichte: 1.32 g/cm³
Flammpunkt: >110 °C
MDL: MFCD00008109
CAS-Nummer: 95-00-1
EINECS: 202-382-6
UN: 2735
ADR: 8 III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5760 to 1.5800 (20°C, 589 nm)
GC ≥97.5 %
Suitability (for use as a laser dye) Passes test
Molar absorption coefficient ≥77000 l/(mol.cm) (at Lambda max)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR