2,4-Dichlorbenzylamin 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
(2,4-Dichlorophenyl)methanamine
113230100
113230500
ACRO113230100EA
37.6
EUR
ACRO113230100
ACRO113230500
2,4-Dichlorbenzylamin 98%
2,4-Dichlorbenzylamin
Formel:
C₇H₇Cl₂N Molecular Weight: 176.04 g/mol Siedepunkt: 83 Dichte: 1.32 g/cm³ Flammpunkt: >110 °C |
MDL:
MFCD00008109 CAS-Nummer: 95-00-1 EINECS: 202-382-6 UN: 2735 ADR: 8 III |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5760 to 1.5800 (20°C, 589 nm) |
GC | ≥97.5 % |
Suitability | (for use as a laser dye) Passes test |
Molar absorption coefficient | ≥77000 l/(mol.cm) (at Lambda max) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...