2,5-Dichlorphenol 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2,5-DCP
113580500
113582500
113580010
ACRO113580500EA
32
EUR
ACRO113580500
ACRO113582500
ACRO113580010
2,5-Dichlorphenol 98%
2,5-Dichlorphenol
Formel:
C₆H₄Cl₂O Siedepunkt: 211 °C (760 mmHg) Schmelzpunkt: 54…58 °C Flammpunkt: 100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002174 CAS-Nummer: 583-78-8 EINECS: 209-520-4 UN: 2020 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to light brown Crystals of flakes |
Infrared spectrum | Conforms |
Melting point | 54 to 58 °C |
GC | ≥97.5 % |
Uniformity coefficient | ≤1.60 |
Moisture holding capacity | 49.0 to 60.0 % (Cl) |
Total volume capacity | ≥1.20 eq/l (Cl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...