(2,4-Dichlorphenoxy)essigsäure 97%

Lieferant: Thermo Fisher Scientific
Gefahr

113631000 113635000
ACRO113631000EA 38 EUR
ACRO113631000 ACRO113635000
(2,4-Dichlorphenoxy)essigsäure 97%
(2,4-Dichlorphenoxy)essigsäure
Formel: (C₆H₃Cl₂)OCH₂COOH
Siedepunkt: 160 °C (,4 mmHg)
Schmelzpunkt: 137…141 °C
Dichte: 1,563 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004300
CAS-Nummer: 94-75-7
EINECS: 202-361-1
Merck Index: 13,02825

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Powder or flakes
Infrared spectrum Conforms
Melting point 136 to 141 °C
HPLC ≥96.0 %
Titration with NaOH 96.0 to 104.0 %
Sulfate (SO4) ≤50 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR