1,2-Diformylhydrazine 97%

Lieferant: Thermo Fisher Scientific

114640250
ACRO114640250EA 170 EUR
ACRO114640250
1,2-Diformylhydrazine 97%
1,2-Diformylhydrazine

Appearance: White to off-white Crystalline powder

Formel: C₂H₄N₂O₂
Schmelzpunkt: 155…160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003275
CAS-Nummer: 628-36-4

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 155 to 160 °C
TLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR