4,4'-Dihydroxybenzophenon 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Bis(4-hydroxyphenyl) ketone

114920050 114920250
ACRO114920050EA 40 EUR
ACRO114920050 ACRO114920250
4,4'-Dihydroxybenzophenon 97%
4,4'-Dihydroxybenzophenon
Formel: C₁₃H₁₀O₃
Molecular Weight: 214.22 g/mol
Siedepunkt: 375
Schmelzpunkt: 217…222 °C
MDL: MFCD00002358
CAS-Nummer: 611-99-4
EINECS: 210-288-1

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
Melting point 217 to 222 °C
GC ≥96.0 %
Titration with NaOH ≥96.0 %

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR