7-(2,3-Dihydroxypropyl)theophyllin 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Dyphylline, 7-(2,3-Dihydroxypropyl)-1,3-dimethyl-1H-purine-2,6(3H,7H)-dione

115051000 115055000
ACRO115051000EA 69.7 EUR
ACRO115051000 ACRO115055000
7-(2,3-Dihydroxypropyl)theophyllin 99%
7-(2,3-Dihydroxypropyl)theophyllin
Formel: C₁₀H₁₄N₄O₄
Molecular Weight: 254.24 g/mol
Schmelzpunkt: 160…165 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005759
CAS-Nummer: 479-18-5
EINECS: 207-526-1

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Infrared spectrum Conforms
Melting point 160 to 165 °C
Loss on drying ≤0.5 % (1 g, 105 °C)
Titration with HClO4 ≥98.5 % (On dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR