7-(2,3-Dihydroxypropyl)theophyllin 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Dyphylline, 7-(2,3-Dihydroxypropyl)-1,3-dimethyl-1H-purine-2,6(3H,7H)-dione
115051000
115055000
ACRO115051000EA
69.7
EUR
ACRO115051000
ACRO115055000
7-(2,3-Dihydroxypropyl)theophyllin 99%
7-(2,3-Dihydroxypropyl)theophyllin
Formel:
C₁₀H₁₄N₄O₄ Molecular Weight: 254.24 g/mol Schmelzpunkt: 160…165 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005759 CAS-Nummer: 479-18-5 EINECS: 207-526-1 |
Spezifikation Testergebnisse
Appearance | White Powder |
Infrared spectrum | Conforms |
Melting point | 160 to 165 °C |
Loss on drying | ≤0.5 % (1 g, 105 °C) |
Titration with HClO4 | ≥98.5 % (On dried substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...