1,2:3,4-Di-O-isopropylidene-ɑ-D-galactopyranose 97%

Lieferant: Thermo Fisher Scientific

115240050 115240250
ACRO115240050EA 40.8 EUR
ACRO115240050 ACRO115240250
1,2:3,4-Di-O-isopropylidene-ɑ-D-galactopyranose 97%
1,2:3,4-Di-O-isopropylidene-alpha-D-galactopyranose

Appearance: Clear colorless to yellow Viscous liquid or paste

Formel: C₁₂H₂₀O₆
Siedepunkt: 117 °C (,1 mmHg)
Schmelzpunkt: 120…122 °C
Dichte: 1,14 g/cm³
Lagertemperatur: Gefrierschrank
MDL: MFCD00063225
CAS-Nummer: 4064-06-6

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Viscous liquid or paste
Infrared spectrum Conforms
Refractive index 1.4655 to 1.4685 (20°C, 589 nm)
GC ≥96.0 %
Specific optical rotation -57° to -61° (20°C, 589 nm) (c=3, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR