4-(Dimethylamino)benzonitril 95%

Lieferant: Thermo Fisher Scientific
Achtung

115780050 115780250
ACRO115780050EA 32.7 EUR
ACRO115780050 ACRO115780250
4-(Dimethylamino)benzonitril 95%
4-(Dimethylamino)benzonitril
Formel: C₉H₁₀N₂
Schmelzpunkt: 70…76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001815
CAS-Nummer: 1197-19-9
EINECS: 214-819-8
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance Light brown Crystalline powder
Infrared spectrum Conforms
Melting point 70 to 76 °C
Titration with HClO4 ≥94 %
Stabilizer About 500 ppm Nitrocresol + Nitrophenol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR