4-(Dimethylamino)benzonitril 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
115780050
115780250
ACRO115780050EA
32.7
EUR
ACRO115780050
ACRO115780250
4-(Dimethylamino)benzonitril 95%
4-(Dimethylamino)benzonitril
Formel:
C₉H₁₀N₂ Schmelzpunkt: 70…76 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001815 CAS-Nummer: 1197-19-9 EINECS: 214-819-8 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 70 to 76 °C |
Titration with HClO4 | ≥94 % |
Stabilizer | About 500 ppm Nitrocresol + Nitrophenol |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...