(2-Chlorpropyl)dimethylammoniumchlorid 98.5%

Lieferant: Thermo Scientific
Achtung

Synonyme: N-(2-Chloropropyl)-N,N-dimethylammonium chloride, 2-Chloro-N,N-dimethylpropan-1-amine hydrochloride, 2-Dimethylaminoisopropyl chloride hydrochloride

115835000
ACRO115835000EA 81.1 EUR
ACRO115835000
(2-Chlorpropyl)dimethylammoniumchlorid 98.5%
2-Chloropropyldimethylammonium chloride
Formel: C₅H₁₃Cl₂N
Molecular Weight: 158,07 g/mol
Schmelzpunkt: 191…195 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012534
CAS-Nummer: 4584-49-0
EINECS: 224-971-7

Bestellen

Spezifikation Testergebnisse

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 %
Titration with HClO4 ≥98.0 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR