3-(Dimethylamino)phenol 97%

Lieferant: Thermo Fisher Scientific
Achtung

115860250 115861000 115865000
ACRO115860250EA 58 EUR
ACRO115860250 ACRO115861000 ACRO115865000
3-(Dimethylamino)phenol 97%
Formel: C₈H₁₁NO
Siedepunkt: 265 °C (1013 hPa)
Schmelzpunkt: 80…85 °C
Flammpunkt: 148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002264
CAS-Nummer: 99-07-0
EINECS: 202-727-0

Bestellen

Spezifikation Testergebnisse

Appearance Grey to brown to purple Powder
Infrared spectrum Conforms
GC Xylene isomers plus Ethylbenzene (Ethylbenzene not to exceed 25%)
Water ≤0.05 % (Coulometric)
Color scale ≤10 APHA
Residue after evaporation ≤0.002 %
Substance darkened by H2SO4 Passes test
Sulfur compounds (as S) ≤0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR