5,6-Dimethylbenzimidazol 99+%

Lieferant: Thermo Fisher Scientific

115980250 115981000
ACRO115980250EA 37.3 EUR
ACRO115980250 ACRO115981000
5,6-Dimethylbenzimidazol 99+%
5,6-Dimethylbenzimidazole
Formel: C₉H₁₀N₂
Siedepunkt: 377 °C (1013 hPa)
Schmelzpunkt: 203…206 °C
MDL: MFCD00005603
CAS-Nummer: 582-60-5
EINECS: 209-488-1
Merck Index: 13,03263

Bestellen

Spezifikation Testergebnisse

Appearance Light beige Powder
Infrared spectrum Conforms
Melting point 203 to 206 °C
Titration with HClO4 ≥99.0 %
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤10 ppm
Magnesium (Mg) ≤20 ppm
Sodium (Na) ≤5000 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR