N,N-Dimethylethylendiamin 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
116201000
116200250
116205000
ACRO116201000EA
68.9
EUR
ACRO116201000
ACRO116200250
ACRO116205000
N,N-Dimethylethylendiamin 99%
N,N-Dimethylethylendiamin
Formel:
C₄H₁₂N₂ Siedepunkt: 105,5…107 °C (760 mmHg) Schmelzpunkt: –70 °C Dichte: 0,8 g/cm³ Flammpunkt: 12 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008175 CAS-Nummer: 108-00-9 EINECS: 203-541-2 UN: 2734 ADR: 8,I |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4255 to 1.4275 (20°C, 589 nm) |
Water | ≤1 % |
GC | ≥98.5 % |
Sodium (Na) | ≤200 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...