3,3-Dimethylglutarsäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3,3-Dimethyl-1,5-pentandisäure

116240250 116241000 116245000
ACRO116240250EA 51.6 EUR
ACRO116240250 ACRO116241000 ACRO116245000
3,3-Dimethylglutarsäure 98%
3,3-Dimethylglutarsäure
Formel: C₇H₁₂O₄
Molecular Weight: 160.17 g/mol
Siedepunkt: 290 °C (760 mmHg)
Schmelzpunkt: 98…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002716
CAS-Nummer: 4839-46-7
EINECS: 225-425-0

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 98 to 102 °C
Water ≤1 % (K.F.)
Titration with NaOH ≥97.5 %
Phosphate (PO4) ≤2 ppm
Sulfate (SO4) ≤2 ppm
Iron (Fe) ≤0.2 ppm
Heavy metals (as Pb) ≤0.5 ppm
Carbon dioxide (CO2) ≤0.002 %
KMnO4-reducing substance Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR