2,6-Xylenol 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2,6-Dimethylphenol, 2-Hydroxy-3-xylol, 2-Hydroxy-m-xylol, sym.-o-Xylenol
116570010
116570050
116572500
116570025
116570051
ACRO116570010EA
79.2
EUR
ACRO116570010
ACRO116570050
ACRO116572500
ACRO116570025
ACRO116570051
2,6-Xylenol 99%
2,6-Xylenol
Formel:
C₈H₁₀O Molecular Weight: 122.17 g/mol Siedepunkt: 203 °C (760 mmHg) Schmelzpunkt: 45…48 °C Dichte: 1.15 g/cm³ Flammpunkt: 73 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002240 CAS-Nummer: 576-26-1 EINECS: 209-400-1 UN: 2261 ADR: 6.1 II Merck Index: 12,10215 |
Spezifikation Testergebnisse
Appearance | Colorless to yellow-brown Low melting solid (May darken on storage) |
Infrared spectrum | Conforms |
Melting point | 45 to 48 °C |
Water | ≤0.05 % |
GC | ≥98.5 % |
Sulfate (SO4) | ≤0.005 % |
Iron (Fe) | ≤3 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.005 % |
Iodine consuming subst. | ≤0.004 meq/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...