Docusatnatrium 96%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Sulfobernsteinsäure-bis-(2-ethylhexylester) Natriumsalz, 1,4-Butandisäuredi(2-ethylhexyl)ester-2-sulfonsäure Natriumsalz

117100010 117101000 117100025
ACRO117100010EA 97.2 EUR
ACRO117100010 ACRO117101000 ACRO117100025
Docusatnatrium 96%
Docusatnatrium
Formel: C₂₀H₃₇NaO₇S
Molecular Weight: 444.57 g/mol
Schmelzpunkt: 153…157 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012455
CAS-Nummer: 577-11-7
EINECS: 209-406-4
Merck Index: 12,03460

Bestellen

Spezifikation Testergebnisse

Appearance White Waxy solid
Infrared spectrum Conforms
Assay ≥95 %
Water ≤2 %
Color of alcohol solution ≤100 APHA (50 % solids in 1:1 alcohol/water)
Insoluble matter ≤300 ppm (50 % in toluene)
Iodine value ≤0.25 (on solids basis)
Acid number ≤2.5 (on solids basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR