1,1-Diphenylethylen 98%

Lieferant: Thermo Fisher Scientific

117260100 117260250 117261000
ACRO117260100EA 51.9 EUR
ACRO117260100 ACRO117260250 ACRO117261000
1,1-Diphenylethylen 98%
1,1-Diphenylethylen
Formel: (C₆H₅)₂CCH₂
Siedepunkt: 270…271 °C (760 mmHg)
Schmelzpunkt: 6 °C
Dichte: 1,02 g/cm³
Flammpunkt: >112 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00008583
CAS-Nummer: 530-48-3
EINECS: 208-482-6
Merck Index: 13,03355
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef9e2a018fc6e0a147414e06d2cdd5b51e949db2&vid=23a75fa0969db1a09c065b9fced36b49bc2d26df&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to golden Liquid
Infrared spectrum Conforms
Refractive index 1.6070 to 1.6090 (20°C, 589 nm)
GC ≥97.5 %
Iodide (I) ≤30 ppm
Phosphate (PO4) ≤10 ppm
SO4 and SO3 (as SO4) ≤0.003 %
Iron (Fe) ≤1 ppm
Selenium (Se) ≤0.01 ppm
Heavy metals (ICP-OES) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR