2,5-Diphenyloxazol 99% zur Szintillation
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
PPO, 2,5-Diphenyl-1,3-oxazol
117380250
117381000
117385000
117380025
ACRO117380250EA
32.8
EUR
ACRO117380250
ACRO117381000
ACRO117385000
ACRO117380025
2,5-Diphenyloxazol 99% zur Szintillation
2,5-Diphenyloxazole
Formel:
C₁₅H₁₁NO Molecular Weight: 221.26 g/mol Siedepunkt: 360 °C (760 mmHg) Schmelzpunkt: 70…72 °C Dichte: 1.06 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005306 CAS-Nummer: 92-71-7 EINECS: 202-181-3 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 70 to 72 °C |
Additional info | background: < 25 cpm |
HPLC | ≥98.5 % |
UV transmittance | ≥85 % (40% in toluene) |
Color of solution | ≤50 APHA (40% in toluene) |
Carbonyl compounds | ≤0.002 % (acetone) |
Titrable acid or base | ≤0.0001 meq/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...