(±)-Epichlorhydrin 99%

Lieferant: Thermo Fisher Scientific
Gefahr

117780010 117780025 117780250 117780100
ACRO117780010EA 66.4 EUR
ACRO117780010 ACRO117780025 ACRO117780250 ACRO117780100
(±)-Epichlorhydrin 99%
(±)-Epichlorhydrin
Formel: C₃H₅ClO
Siedepunkt: 115…117 °C (760 mmHg)
Schmelzpunkt: –57 °C
Dichte: 1,18 g/cm³
Flammpunkt: 28 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00005132
CAS-Nummer: 106-89-8
EINECS: 203-439-8
UN: 2023
ADR: 6.1,II
Merck Index: 13,03642

Bestellen

Spezifikation Testergebnisse

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4365 to 1.4385 (20°C, 589 nm)
Water ≤0.02 % (K.F.)
GC ≥98.5 %
Color scale ≤15 APHA
Silicate and phosphate ≤5 ppm (as SiO2)
Iron (Fe) ≤1 ppm
Heavy metals (as Pb) ≤1 ppm
Nitrogen compounds (as N) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR