1,2-Ethandithiol 95%

Lieferant: Thermo Fisher Scientific
Gefahr

117852500 117850010
ACRO117852500EA 92.1 EUR
ACRO117852500 ACRO117850010
1,2-Ethandithiol 95%
1,2-Ethandithiol
Formel: C₂H₆S₂
Siedepunkt: 144…146 °C (760 mmHg)
Schmelzpunkt: –41 °C
Dichte: 1,123 g/cm³
Flammpunkt: 50 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004892
CAS-Nummer: 540-63-6
EINECS: 208-752-3
UN: 3071
ADR: 6.1,II
Merck Index: 13,03760

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow to slightly colored Liquid
Infrared spectrum Conforms
GC ≥94 %
Bromide (Br) ≤0.01 %
Iodide (I) ≤20 ppm
Chlorate and Nitrate (as NO3) ≤0.003 %
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤10 ppm
Barium (Ba) Passes test (Limit about 0.001%)
Calcium (Ca) ≤20 ppm
Iron (Fe) ≤3 ppm
Magnesium (Mg) ≤10 ppm
Sodium (Na) ≤50 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR