Ethyl-3-aminobenzoat 99+%

Lieferant: Thermo Fisher Scientific
Achtung

117990050 117990250
ACRO117990050EA 28.3 EUR
ACRO117990050 ACRO117990250
Ethyl-3-aminobenzoat 99+%
Ethyl-3-aminobenzoat
Formel: C₉H₁₁NO₂
Siedepunkt: 172…175 °C (13 mmHg)
Dichte: 1,107 g/cm³
Flammpunkt: >112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007794
CAS-Nummer: 582-33-2
EINECS: 209-482-9

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow to light brown Liquid or low melting solid
Infrared spectrum Conforms
Refractive index 1.5595 to 1.5615 (20°C, 589 nm)
GC ≥99.0 %
Chloride (Cl) ≤10 ppm
Calcium (Ca) ≤0.01 %
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤0.002 %
Sodium (Na) ≤0.02 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR