(±)-Ethyl-2-brompropionat 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
DL-Ethyl 2-bromopropionate, Ethyl 2-bromopropionate
118141000
118142500
118140010
ACRO118141000EA
39.8
EUR
ACRO118141000
ACRO118142500
ACRO118140010
(±)-Ethyl-2-brompropionat 99%
(±)-Ethyl-2-brompropionat
Formel:
C₅H₉BrO₂ Molecular Weight: 181.03 g/mol Siedepunkt: 156…160 °C (760 mmHg) Dichte: 1.394 g/cm³ Flammpunkt: 51 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000144 CAS-Nummer: 535-11-5 EINECS: 208-609-5 UN: 2920 ADR: 8 II |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4450 to 1.4470 (20°C, 589 nm) |
GC | ≥98.5 % |
Sulfate (SO4) | ≤300 ppm |
Sodium fluosilicate | ≤0.1 % (Na2SiF6) |
Sulfite (SO2) | ≤50 ppm |
Iron (Fe) | ≤30 ppm |
Potassium (K) | ≤200 ppm |
Heavy metals (as Pb) | ≤30 ppm |
Titrable acid | ≤0.03 meq/g |
Titrable base | ≤0.01 meq/g |
Insoluble matter | ≤0.02 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...