Ethoxymethylen-cyanessigsäureethylester 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
118481000
118485000
118480010
ACRO118481000EA
40.5
EUR
ACRO118481000
ACRO118485000
ACRO118480010
Ethoxymethylen-cyanessigsäureethylester 98%
Ethoxymethylen-cyanessigsäureethylester
Formel:
C₈H₁₁NO₃ Siedepunkt: 190…191 °C (30 mmHg) Schmelzpunkt: 48…52 °C Flammpunkt: 130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009136 CAS-Nummer: 94-05-3 EINECS: 202-299-5 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to yellow to yellow-brown Powder or crystals or flakes or crystalline solid |
Infrared spectrum | Conforms |
Melting point | 47 to 53 °C |
GC | ≥99.5 % |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.001 % |
Substance darkened by H2SO4 | passes test |
Sulfur compounds (as S) | ≤0.003 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...