Thiomersal 97,0-101,0%

Lieferant: Thermo Fisher Scientific
Gefahr

118680050 118680250 118681000 118685000
ACRO118680050EA 115 EUR
ACRO118680050 ACRO118680250 ACRO118681000 ACRO118685000
Thiomersal 97,0-101,0%
Thiomersal
Formel: 2-(C₂H₅HgS)C₆H₄CO₂Na
Flammpunkt: 250 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013062
CAS-Nummer: 54-64-8
EINECS: 200-210-4
UN: 2025
ADR: 6.1,II
Merck Index: 12,09451
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=621fb9ecf6f0ee8129acf49c938a6167645828da&vid=a4d9481c20c7c69ffe3a642fbe6cae8c3dfb117f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White to cream Powder
Infrared spectrum Conforms
Assay 97.0 to 101.0 % (On dried substance)
Loss on drying ≤0.5 % (vacuum/P2O5)
Additional info ion mercuric: ≤ 0.7%
pH 6 to 8 (1 % solution)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR