Ethylphenylcyanacetat 97%
Lieferant: Thermo Scientific
|
Achtung
|
118830050
ACRO118830050EA
39.9
EUR
ACRO118830050
Ethylphenylcyanacetat 97%
Ethylphenylcyanacetat
Formel:
C₁₁H₁₁NO₂ Molecular Weight: 189,21 g/mol Dichte: 1,09 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001866 CAS-Nummer: 4553-07-5 EINECS: 224-921-4 UN: 2810 ADR: 6.1,II |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5033 to 1.5053 (20°C, 589 nm) |
Titration with NaOH | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...