Piperidin-N-carbaldehyd 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
119730010
119732500
ACRO119730010EA
277
EUR
ACRO119730010
ACRO119732500
Piperidin-N-carbaldehyd 99%
Piperidin-N-carbaldehyd
Formel:
C₆H₁₁NO Schmelzpunkt: –31 °C Dichte: 1,019 g/cm³ Flammpunkt: 91 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006483 CAS-Nummer: 2591-86-8 EINECS: 219-986-0 UN: 2810 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4830 to 1.4850 (20°C, 589 nm) |
GC | ≥98.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...