Girard-Reagenz T (Carbazoylmethyltrimethylammoniumchlorid) 99%

Lieferant: Thermo Fisher Scientific
Achtung

119870500 119872500
ACRO119870500EA 34.2 EUR
ACRO119870500 ACRO119872500
Girard-Reagenz T (Carbazoylmethyltrimethylammoniumchlorid) 99%
Girard-Reagenz T (Carbazoylmethyltrimethylammoniumchlorid)
Formel: C₅H₁₄ClN₃O
Schmelzpunkt: 188…192 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012009
CAS-Nummer: 123-46-6
EINECS: 204-629-3

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 187 to 192 °C
Titration Argentometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR