DL-Homocysteinthiolactonhydrochlorid 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
DL-3-Aminotetrahydrothiophen-2-one Hydrochloride, 3-Aminodihydrothiophen-2(3H)-one hydrochloride, DL-Homocysteine thiolactone HCl
120830250
120831000
ACRO120830250EA
27.5
EUR
ACRO120830250
ACRO120831000
DL-Homocysteinthiolactonhydrochlorid 99%
DL-Homocysteinethiolactone hydrochloride
Appearance: White Crystalline powder
Formel:
C₄H₇NOS Schmelzpunkt: 199…203 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012724 CAS-Nummer: 6038-19-3 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...