DL-Homocysteinthiolactonhydrochlorid 99%

Lieferant: Thermo Fisher Scientific

Synonyme: DL-3-Aminotetrahydrothiophen-2-one Hydrochloride, 3-Aminodihydrothiophen-2(3H)-one hydrochloride, DL-Homocysteine thiolactone HCl

120830250 120831000
ACRO120830250EA 27.5 EUR
ACRO120830250 ACRO120831000
DL-Homocysteinthiolactonhydrochlorid 99%
DL-Homocysteinethiolactone hydrochloride

Appearance: White Crystalline powder

Formel: C₄H₇NOS
Molecular Weight: 153.62 g/mol
Schmelzpunkt: 199…203 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012724
CAS-Nummer: 6038-19-3

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR