trans-m-Coumaric acid 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: (2E)-3-(3-Hydroxyphenyl)acrylic acid, m-Hydroxycinnamic acid, predominantly trans, trans-3-Hydroxycinnamic acid, (E)-m-Coumaric acid, (E)-3-(3-Hydroxyphenyl)acrylic acid

121080250
ACRO121080250EA 49.7 EUR
ACRO121080250
trans-m-Coumaric acid 99%
trans-m-Coumaric acid

  • Predominantly trans

Formel: C₉H₈O₃
Molecular Weight: 164,16 g/mol
Schmelzpunkt: 189…192 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004390
CAS-Nummer: 14755-02-3
EINECS: 209-615-0

Bestellen

Spezifikation Testergebnisse

Appearance Beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Specific optical rotation +12° to +16° (20°C, 589 nm) (c=1, ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR