9-Hydroxyfluoren 97%
Lieferant: Thermo Scientific
Synonyme:
9-Fluorenol
121230250
ACRO121230250EA
56
EUR
ACRO121230250
9-Hydroxyfluoren 97%
9-Hydroxyfluoren
Formel:
C₁₃H₁₀O Molecular Weight: 182,22 g/mol Siedepunkt: 320 °C (1013 hPa) Schmelzpunkt: 152…158 °C Dichte: 1,151 g/cm³ (20 °C) |
MDL:
MFCD00001135 CAS-Nummer: 1689-64-1 EINECS: 216-879-0 |
Spezifikation Testergebnisse
Appearance | White to cream Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 152 to 158 °C |
GC | ≥96.0 % |
Titration with HClO4 | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...