5-Hydroxy-3-indolylessigsäure 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
5-Hydroxyindol-3-essigsäure, 5-Hydroxyindol-3-ylessigsäure
121280010
121282500
121280050
ACRO121280010EA
202
EUR
ACRO121280010
ACRO121282500
ACRO121280050
5-Hydroxy-3-indolylessigsäure 99%
5-Hydroxy-3-indolylessigsäure
Formel:
C₁₀H₉NO₃ Schmelzpunkt: 162…165 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00005639 CAS-Nummer: 54-16-0 EINECS: 200-195-4 |
Spezifikation Testergebnisse
Appearance | Beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 162 to 165 °C |
Water | ≤0.5 % (K.F.) |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...