4-Hydroxy-3-methoxybenzylamin Hydrochlorid 98%

Lieferant: Thermo Fisher Scientific
Achtung

121370010 121370050
ACRO121370010EA 27.2 EUR
ACRO121370010 ACRO121370050
4-Hydroxy-3-methoxybenzylamin Hydrochlorid 98%
4-Hydroxy-3-methoxybenzylamin Hydrochlorid
Formel: C₈H₁₂ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00012864
CAS-Nummer: 7149-10-2
EINECS: 230-468-3

Bestellen

Spezifikation Testergebnisse

Appearance Off-white Powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR