N-(Hydroxymethyl)phthalimid 97%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-(Hydroxymethyl)phthalsäureimid, N-(Hydroxymethyl)-1,2-benzoldicarbonsäureimid
121490250
121491000
ACRO121490250EA
25.9
EUR
ACRO121490250
ACRO121491000
N-(Hydroxymethyl)phthalimid 97%
N-(Hydroxymethyl)phthalimid
Formel:
C₉H₇NO₃ Siedepunkt: 415 °C (1013 hPa) Schmelzpunkt: 140…145 °C |
MDL:
MFCD00005899 CAS-Nummer: 118-29-6 EINECS: 204-241-4 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 140 to 145 °C |
Loss on drying | ≤2 % |
Sulfated ash | ≤0.1 % |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...