(4-Hydroxyphenyl)essigsäure 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Hydroxyphenylacetic Acid
121710250
121711000
ACRO121710250EA
60
EUR
ACRO121710250
ACRO121711000
(4-Hydroxyphenyl)essigsäure 98%
(4-Hydroxyphenyl)essigsäure
Formel:
(C₆H₄(OH))CH₂COOH Molecular Weight: 152.15 g/mol Siedepunkt: 347 Schmelzpunkt: 147…152 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004347 CAS-Nummer: 156-38-7 EINECS: 205-851-3 |
Spezifikation Testergebnisse
Appearance | White to cream to tan Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 147 to 152 °C |
Water | ≤0.5 % (K.F.) |
TLC | ≤1 % Methyl ester derivative |
Titration with NaOH | 97.5 to 102.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...