(4-Hydroxyphenyl)essigsäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Hydroxyphenylacetic Acid

121710250 121711000
ACRO121710250EA 60 EUR
ACRO121710250 ACRO121711000
(4-Hydroxyphenyl)essigsäure 98%
(4-Hydroxyphenyl)essigsäure
Formel: (C₆H₄(OH))CH₂COOH
Molecular Weight: 152.15 g/mol
Siedepunkt: 347
Schmelzpunkt: 147…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004347
CAS-Nummer: 156-38-7
EINECS: 205-851-3

Bestellen

Spezifikation Testergebnisse

Appearance White to cream to tan Crystalline powder
Infrared spectrum Conforms
Melting point 147 to 152 °C
Water ≤0.5 % (K.F.)
TLC ≤1 % Methyl ester derivative
Titration with NaOH 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR