trans-4-Hydroxy-L(-)-prolin 99+%

Lieferant: Thermo Fisher Scientific

Synonyme: L(-)-trans-4-Hydroxyprolin, (S)-(-)-trans-4-Hydroxyprolin, H-Hyp-OH, (2S)-(-)-trans-4-Hydroxypyrrolidin-2-carbonsäure, (S)-(-)-trans-4-Hydroxy-1,3,4,5-tetrahydro-2H-pyrrol- 2-carbonsäure, (S)-(-)-trans-4-Hydroxyazolidin-2-carbonsäure, (S)-(-)-trans-4-Hydroxypyrrolidin-2-carbonsäure

121780100 121780250 121781000 121785000
ACRO121780100EA 64.1 EUR
ACRO121780100 ACRO121780250 ACRO121781000 ACRO121785000
trans-4-Hydroxy-L(-)-prolin 99+%
trans-4-Hydroxy-L(-)-prolin
Formel: C₅H₉NO₃
Siedepunkt: 296 °C (1013 hPa)
Schmelzpunkt: 274…275 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064320
CAS-Nummer: 51-35-4
EINECS: 200-091-9
Merck Index: 13,04866

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.2 % (105°C, 3 hrs)
Sulfated ash ≤0.1 %
UV transmittance ≥95 % (c=10, H2O, 430 nm) 1 cm cell
Specific optical rotation -74° to -77° (20°C, 589 nm) (c=5, H2O)
Titration with HClO4 99.0 to 101.0 %
Chloride (Cl) ≤200 ppm
Sulfate (SO4) ≤200 ppm
Ammonium (NH4) ≤200 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤10 ppm
Other amino acids ≤0.5 % (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR