Inosin 99%

Lieferant: Thermo Fisher Scientific

Synonyme: 9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one

122250100 122250250 122251000 122252500
ACRO122250100EA 63.6 EUR
ACRO122250100 ACRO122250250 ACRO122251000 ACRO122252500
Inosin 99%
Inosin
Formel: C₁₀H₁₂N₄O₅
Molecular Weight: 268.23 g/mol
Schmelzpunkt: 212…213 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00066770
CAS-Nummer: 58-63-9
EINECS: 200-390-4

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 %
Sulfated ash ≤0.1 %
HPLC ≥98.5 %
UV spectrophotometry Molar absorptivity 12200 +/- 2%, on dry substance
UV transmittance ≥98 % (c=1, H2O, 430 nm, 1 cm cell)
Ratio of Absorbances 0.22 to 0.26 (A280/A260)
Titration with HClO4 ≥98.5 % (On dry substance)
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR