5-Iod-2'-deoxyuridin (Idoxuridin) 99%

Lieferant: Thermo Fisher Scientific
Achtung

122350010 122350050 122350250
ACRO122350010EA 54.2 EUR
ACRO122350010 ACRO122350050 ACRO122350250
5-Iod-2'-deoxyuridin (Idoxuridin) 99%
5-Iod-2'-deoxyuridin (Idoxuridin)
Formel: C₉H₁₁IN₂O₅
Schmelzpunkt: 155…180 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00134656
CAS-Nummer: 54-42-2
EINECS: 200-207-8

Bestellen

Spezifikation Testergebnisse

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 % (1 g, 60°C) (vacuum)
Specific optical rotation +28° to +32° (20°C, 589 nm) (c=1, 1 N NaOH) (on dried substance)
Titration with NaOH ≥98.5 % (On dried substance)
Oxalate (C2O4) (limit about 0.05%) Passes test
Phosphate (PO4) ≤0.001 %
Sulfate (SO4) ≤0.002 %
Iron (Fe) ≤3 ppm
Lead (Pb) ≤2 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR