2-(4-Iodphenyl)-3-(4-nitrophenyl)-5-phenyltetrazoliumchlorid 98%

Lieferant: Thermo Fisher Scientific
Achtung

122400010 122400050 122400250
ACRO122400010EA 80.2 EUR
ACRO122400010 ACRO122400050 ACRO122400250
2-(4-Iodphenyl)-3-(4-nitrophenyl)-5-phenyltetrazoliumchlorid 98%
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride
Formel: C₁₉H₁₃ClIN₅O₂
Lagertemperatur: Kühlschrank
MDL: MFCD00011961
CAS-Nummer: 146-68-9
EINECS: 205-676-2

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
GC ≥98.5 %
NMR (1H-NMR) Consistent with structure
Specific optical rotation +6.3° to +7.3° (20°C, 589 nm) (C=4, water) (Warm at 40°C to dissolve)
Titration with HClO4 98.5 to 101.5 % (On dried basis)
Chloride (Cl) ≤0.05 %
Sulfate (SO4) ≤0.03 %
Iron (Fe) ≤30 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR