6,7-Dihydrobenzo[b]thiophen-4-one 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Oxo-4,5,6,7-tetrahydrothianaphthene, 6,7-Dihydro-4-benzo[b]thiophenone, 4-Keto-4,5,6,7-tetrahydrothianaphthene

122890010
ACRO122890010EA 68.9 EUR
ACRO122890010
6,7-Dihydrobenzo[b]thiophen-4-one 97%
6,7-Dihydrobenzo[b]thiophen-4-one

Appearance: White to yellow Low melting solid or liquid after melting

Formel: C₈H₈OS
Molecular Weight: 152,211 g/mol
Siedepunkt: 120 °C (10 mmHg)
Flammpunkt: 110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005861
CAS-Nummer: 13414-95-4

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Low melting solid or liquid after melting
Infrared spectrum Conforms
Melting point 34 to 40 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR