Natriumacetat Trihydrat 99+%, reinst
Lieferant: Thermo Fisher Scientific
123240010
123240050
ACRO123240010EA
54.4
EUR
ACRO123240010
ACRO123240050
Natriumacetat Trihydrat 99+%, reinst
Natriumacetat Trihydrat
Formel:
H₃CCOONa·3H₂O Schmelzpunkt: 58 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00071557 CAS-Nummer: 6131-90-4 EINECS: 204-823-8 Merck Index: 13,08642 |
|
Spezifikation Testergebnisse
Appearance | White Adhering crystals |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥99.0 % |
Chloride (Cl) | ≤200 ppm |
Sulfate (SO4) | ≤200 ppm |
Iron (Fe) | ≤10 ppm |
Potassium (K) | ≤500 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Insoluble matter | ≤0.05 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...