1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid 97%

Lieferant: Thermo Fisher Scientific
Achtung

123310010 123310050 123311000
ACRO123310010EA 106 EUR
ACRO123310010 ACRO123310050 ACRO123311000
1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid 97%
1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid
Formel: C₂₂H₂₄N₂O₁₀
Schmelzpunkt: 177…179 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00036255
CAS-Nummer: 85233-19-8

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR