Natriumthiosulfat solution_standard 0.1 mol/l (0.1 N) water

Lieferant: Thermo Fisher Scientific

Synonyme: di-Natriumthiosulfat, Antichlor, Fixiersalz, Natriumhyposulfit

124270010 124270025 124270250
ACRO124270010EA 38.7 EUR
ACRO124270010 ACRO124270025 ACRO124270250
Natriumthiosulfat solution_standard 0.1 mol/l (0.1 N) water
Natriumthiosulfat
Formel: Na₂S₂O₃
Molecular Weight: 158.11 g/mol
Siedepunkt: 100
Schmelzpunkt: 48
Lagertemperatur: Raumtemperatur
MDL: MFCD00003499
CAS-Nummer: 7772-98-7
EINECS: 231-867-5
Merck Index: 12,08844

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Solution
Titration Iodimetric 0.0950 to 0.1050 N (20°C)
Assay ≥90 % (DAC)
Loss on drying ≤10.0 %
Molecular weight 600000 to 800000 Da
Viscosity 50 to 200 mPa.s
Ash ≤1.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR